CH4102152864 ChemBridge Corporation

1-(chloroacetyl)-6-methyl-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD08691894
  • Cas Number : 57368-83-9
  • Molecular Weight : 223.6987

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070668

Safety Information

MSDS https://www.hit2lead.com/sds/9070668_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart