CH4102152883 ChemBridge Corporation

3-sec-butoxybenzaldehyde

  • MDL number MFCD08691916
  • Cas Number : 915924-09-3
  • Molecular Weight : 178.2277

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071037

Safety Information

MSDS https://www.hit2lead.com/sds/9071037_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart