CH4102152914 ChemBridge Corporation

2-(ethylthio)-1,3-benzoxazol-6-amine

  • MDL number MFCD08691959
  • Cas Number : 874463-07-7
  • Molecular Weight : 194.2535

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071503

Safety Information

MSDS https://www.hit2lead.com/sds/9071503_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart