CH4102152952 ChemBridge Corporation

5-bromo-N-methyl-2-furamide

  • MDL number MFCD06093352
  • Cas Number : 31136-78-4
  • Molecular Weight : 204.0213

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7344619

Safety Information

MSDS https://www.hit2lead.com/sds/7344619_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart