CH4102152955 ChemBridge Corporation

N-(4-methylbenzyl)-3-oxobutanamide

  • MDL number MFCD03584671
  • Cas Number : 710307-99-6
  • Molecular Weight : 205.253

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7361334

Safety Information

MSDS https://www.hit2lead.com/sds/7361334_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart