CH4102152956 ChemBridge Corporation

2-isobutoxyphenol

  • MDL number MFCD00474627
  • Cas Number : 21315-20-8
  • Molecular Weight : 166.217

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7365290

Safety Information

MSDS https://www.hit2lead.com/sds/7365290_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart