CH4102152960 ChemBridge Corporation

isopropyl 3-(2-amino-4-hydroxy-6-methyl-5-pyrimidinyl)propanoate

  • MDL number MFCD03300451
  • Cas Number : 497228-78-1
  • Molecular Weight : 239.271

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7396087

Safety Information

MSDS https://www.hit2lead.com/sds/7396087_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart