CH4102152976 ChemBridge Corporation

5-(4-ethoxyphenyl)-2-furaldehyde

  • MDL number MFCD00857679
  • Cas Number : 110360-10-6
  • Molecular Weight : 216.2326

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7515555

Safety Information

MSDS https://www.hit2lead.com/sds/7515555_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart