CH4102152985 ChemBridge Corporation

N-ethyl-N-(2-hydroxy-5-methylphenyl)propanamide

  • MDL number MFCD08583148
  • Cas Number : 909361-84-8
  • Molecular Weight : 207.2689

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7638260

Safety Information

MSDS https://www.hit2lead.com/sds/7638260_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart