CH4102152992 ChemBridge Corporation

3-amino-4-(3-methyl-1-piperidinyl)benzamide

  • MDL number MFCD08691781
  • Cas Number : 915920-42-2
  • Molecular Weight : 233.3095

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7669908

Safety Information

MSDS https://www.hit2lead.com/sds/7669908_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart