CH4102153012 ChemBridge Corporation

isopropyl (4-formylphenoxy)acetate

  • MDL number MFCD04327989
  • Cas Number : 199177-25-8
  • Molecular Weight : 222.2372

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7731199

Safety Information

MSDS https://www.hit2lead.com/sds/7731199_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart