CH4102153013 ChemBridge Corporation

4-(acetylamino)-2-chlorobenzoic acid

  • MDL number MFCD03756645
  • Cas Number : 38667-55-9
  • Molecular Weight : 213.6177

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7732073

Safety Information

MSDS https://www.hit2lead.com/sds/7732073_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart