CH4102153018 ChemBridge Corporation

4-sec-butoxy-3-chloro-5-methoxybenzaldehyde

  • MDL number MFCD02629670
  • Cas Number : 872183-59-0
  • Molecular Weight : 242.6987

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6539175

Safety Information

MSDS https://www.hit2lead.com/sds/6539175_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart