CH4102153034 ChemBridge Corporation

2-[(methoxyacetyl)amino]benzoic acid

  • MDL number MFCD03784903
  • Cas Number : 215102-53-7
  • Molecular Weight : 209.1986

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7784008

Safety Information

MSDS https://www.hit2lead.com/sds/7784008_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart