CH4102153039 ChemBridge Corporation

2-(4-formylphenoxy)-N-1,3-thiazol-2-ylacetamide

  • MDL number MFCD08575597
  • Cas Number : 215503-93-8
  • Molecular Weight : 262.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7865702

Safety Information

MSDS https://www.hit2lead.com/sds/7865702_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart