CH4102153047 ChemBridge Corporation

5-[(2,3-dimethylphenoxy)methyl]-1,3,4-thiadiazol-2-amine

  • MDL number MFCD01346791
  • Cas Number : 123217-00-5
  • Molecular Weight : 235.3054

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6627526

Safety Information

MSDS https://www.hit2lead.com/sds/6627526_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart