CH4102153054 ChemBridge Corporation

3-[3-(2-fluorophenyl)-1,2,4-oxadiazol-5-yl]propanoic acid

  • MDL number MFCD05031219
  • Cas Number : 685525-40-0
  • Molecular Weight : 236.1992

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7929547

Safety Information

MSDS https://www.hit2lead.com/sds/7929547_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart