CH4102153058 ChemBridge Corporation

4-[3-(4-methylphenyl)-1,2,4-oxadiazol-5-yl]butanoic acid

  • MDL number MFCD06383511
  • Cas Number : 851628-34-7
  • Molecular Weight : 246.2619

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7939500

Safety Information

MSDS https://www.hit2lead.com/sds/7939500_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart