CH4102153066 ChemBridge Corporation

3-methoxy-2-[(2-methyl-2-propen-1-yl)oxy]benzaldehyde

  • MDL number MFCD08691807
  • Cas Number : 110124-13-5
  • Molecular Weight : 206.2378

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7965191

Safety Information

MSDS https://www.hit2lead.com/sds/7965191_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart