CH4102153075 ChemBridge Corporation

N-(4-hydroxyphenyl)-5-methyl-2-furamide

  • MDL number MFCD08691813
  • Cas Number : 915922-65-5
  • Molecular Weight : 217.2206

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7994587

Safety Information

MSDS https://www.hit2lead.com/sds/7994587_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart