CH4102153077 ChemBridge Corporation

5-(3,4-dihydro-2(1H)-isoquinolinylmethyl)-2-furoic acid

  • MDL number MFCD08691815
  • Cas Number : 915922-68-8
  • Molecular Weight : 257.2845

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7995737

Safety Information

MSDS https://www.hit2lead.com/sds/7995737_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart