CH4102153083 ChemBridge Corporation

5-[(2,4-dimethylphenoxy)methyl]-1,3,4-thiadiazol-2-amine

  • MDL number MFCD01346792
  • Cas Number : 119869-04-4
  • Molecular Weight : 235.3054

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6632710

Safety Information

MSDS https://www.hit2lead.com/sds/6632710_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart