CH4102153099 ChemBridge Corporation

2-sec-butoxybenzaldehyde

  • MDL number MFCD02629688
  • Cas Number : 22921-59-1
  • Molecular Weight : 178.2277

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6539813

Safety Information

MSDS https://www.hit2lead.com/sds/6539813_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart