CH4102153139 ChemBridge Corporation

1-(3,5-dichloro-1H-1,2,4-triazol-1-yl)acetone

  • MDL number MFCD03061431
  • Cas Number : 625401-77-6
  • Molecular Weight : 194.0187

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6661142

Safety Information

MSDS https://www.hit2lead.com/sds/6661142_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart