CH4102153146 ChemBridge Corporation

2-(4-aminophenyl)-4-quinolinecarboxamide

  • MDL number MFCD02602287
  • Cas Number : 444151-71-7
  • Molecular Weight : 263.2939

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6667419

Safety Information

MSDS https://www.hit2lead.com/sds/6667419_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart