CH4102153158 ChemBridge Corporation

4-(3-pyridin-3-yl-1,2,4-oxadiazol-5-yl)butanoic acid

  • MDL number MFCD07186540
  • Cas Number : 878437-11-7
  • Molecular Weight : 233.2233

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9033151

Safety Information

MSDS https://www.hit2lead.com/sds/9033151_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart