CH4102153164 ChemBridge Corporation

N-(4-aminophenyl)-3-phenylpropanamide

  • MDL number MFCD07072454
  • Cas Number : 886713-07-1
  • Molecular Weight : 240.3003

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9035453

Safety Information

MSDS https://www.hit2lead.com/building-blocks/9035453
Availability details for
Quantity
  • Availability
  • Add to Cart