CH4102153176 ChemBridge Corporation

(3-methyl-2-oxo-3,4-dihydroquinazolin-1(2H)-yl)acetic acid

  • MDL number MFCD05257419
  • Cas Number : 696637-93-1
  • Molecular Weight : 220.2246

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9039842

Safety Information

MSDS https://www.hit2lead.com/sds/9039842_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart