CH4102153180 ChemBridge Corporation

3-[3-(2-furyl)-1,2,4-oxadiazol-5-yl]propanoic acid

  • MDL number MFCD07186541
  • Cas Number : 878437-14-0
  • Molecular Weight : 208.1708

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9040954

Safety Information

MSDS https://www.hit2lead.com/sds/9040954_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart