CH4102153184 ChemBridge Corporation

N-(3-aminophenyl)-2-phenylacetamide

  • MDL number MFCD01806275
  • Cas Number : 85856-32-2
  • Molecular Weight : 226.2738

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9041581

Safety Information

MSDS https://www.hit2lead.com/building-blocks/9041581
Availability details for
Quantity
  • Availability
  • Add to Cart