CH4102153191 ChemBridge Corporation

3-chloro-4-(2-methyl-1-piperidinyl)aniline

  • MDL number MFCD07365134
  • Cas Number : 893750-67-9
  • Molecular Weight : 224.7298

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9068513

Safety Information

MSDS https://www.hit2lead.com/building-blocks/9068513
Availability details for
Quantity
  • Availability
  • Add to Cart