CH4102153192 ChemBridge Corporation

5-chloro-2-(3-methyl-1-piperidinyl)aniline

  • MDL number MFCD07365171
  • Cas Number : 893751-41-2
  • Molecular Weight : 224.7298

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9069148

Safety Information

MSDS https://www.hit2lead.com/sds/9069148_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart