CH4102153214 ChemBridge Corporation

3-{[(5-methyl-2-thienyl)methyl]amino}benzoic acid

  • MDL number MFCD07391543
  • Cas Number : 869950-50-5
  • Molecular Weight : 247.3128

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070461

Safety Information

MSDS https://www.hit2lead.com/sds/9070461_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart