CH4102153225 ChemBridge Corporation

(4-ethoxyphenyl)(2-furyl)methanone

  • MDL number MFCD00094249
  • Cas Number : 21493-98-1
  • Molecular Weight : 216.2326

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070486

Safety Information

MSDS https://www.hit2lead.com/sds/9070486_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart