CH4102153228 ChemBridge Corporation

3-chloro-N-(4-fluoro-2-methylphenyl)propanamide

  • MDL number MFCD08077757
  • Cas Number : 895664-33-2
  • Molecular Weight : 215.6518

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070489

Safety Information

MSDS https://www.hit2lead.com/sds/9070489_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart