CH4102153231 ChemBridge Corporation

2-(2-allylphenoxy)-N-methylethanamine

  • MDL number MFCD08691874
  • Cas Number : 57162-94-4
  • Molecular Weight : 191.2695

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070498

Safety Information

MSDS https://www.hit2lead.com/sds/9070498_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart