CH4102153234 ChemBridge Corporation

4-(4-methoxyphenyl)-5-propyl-1,3-thiazol-2-amine

  • MDL number MFCD03249324
  • Cas Number : 461436-76-0
  • Molecular Weight : 248.3439

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6878082

Safety Information

MSDS https://www.hit2lead.com/sds/6878082_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart