CH4102153262 ChemBridge Corporation

N-methyl-3-(4-methylphenoxy)-1-propanamine

  • MDL number MFCD08691882
  • Cas Number : 915923-08-9
  • Molecular Weight : 179.2588

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070580

Safety Information

MSDS https://www.hit2lead.com/sds/9070580_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart