CH4102153294 ChemBridge Corporation

methyl 5-{[(chloroacetyl)amino]methyl}-2-furoate

  • MDL number MFCD02029657
  • Cas Number : 345991-81-3
  • Molecular Weight : 231.633

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6974944

Safety Information

MSDS https://www.hit2lead.com/sds/6974944_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart