CH4102153299 ChemBridge Corporation

5-(2-chlorophenyl)-2-methyl-3-furoic acid

  • MDL number MFCD08691895
  • Cas Number : 4414-49-7
  • Molecular Weight : 236.6511

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070679

Safety Information

MSDS https://www.hit2lead.com/sds/9070679_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart