CH4102153340 ChemBridge Corporation

5-[(2-hydroxyphenyl)amino]-6-methyl-1,2,4-triazin-3(2H)-one

  • MDL number MFCD20503048
  • Cas Number : 685551-55-7
  • Molecular Weight : 218.212

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070797

Safety Information

MSDS https://www.hit2lead.com/sds/9070797_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart