CH4102153353 ChemBridge Corporation

5-methoxy-8-methyl-3,4-dihydro-1(2H)-naphthalenone

  • MDL number MFCD00820258
  • Cas Number : 53863-68-6
  • Molecular Weight : 190.2384

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070835

Safety Information

MSDS https://www.hit2lead.com/sds/9070835_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart