CH4102153365 ChemBridge Corporation

3-(isobutyrylamino)-4-methylbenzoic acid

  • MDL number MFCD08569891
  • Cas Number : 915921-46-9
  • Molecular Weight : 221.2524

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070873

Safety Information

MSDS https://www.hit2lead.com/sds/9070873_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart