CH4102153384 ChemBridge Corporation

3-[3-(3-methylphenyl)-1,2,4-oxadiazol-5-yl]propanoic acid

  • MDL number MFCD07186380
  • Cas Number : 94192-16-2
  • Molecular Weight : 232.2353

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070924

Safety Information

MSDS https://www.hit2lead.com/sds/9070924_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart