CH4102153391 ChemBridge Corporation

(2-furylmethyl)propylamine

  • MDL number MFCD07410480
  • Cas Number : 39191-12-3
  • Molecular Weight : 139.1949

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070945

Safety Information

MSDS https://www.hit2lead.com/sds/9070945_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart