CH4102153425 ChemBridge Corporation

3-[(2-methylbenzyl)amino]benzoic acid

  • MDL number MFCD08691914
  • Cas Number : 915923-22-7
  • Molecular Weight : 241.2851

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071022

Safety Information

MSDS https://www.hit2lead.com/sds/9071022_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart