CH4102153472 ChemBridge Corporation

2-amino-6-ethyl-5-methyl[1,2,4]triazolo[1,5-a]pyrimidin-7-ol

  • MDL number MFCD00461179
  • Cas Number : 392315-45-6
  • Molecular Weight : 193.2058

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071140

Safety Information

MSDS https://www.hit2lead.com/sds/9071140_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart