CH4102153479 ChemBridge Corporation

2-[(2-furylmethyl)amino]-1-butanol

  • MDL number MFCD07410477
  • Cas Number : 892582-00-2
  • Molecular Weight : 169.2209

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071151

Safety Information

MSDS https://www.hit2lead.com/sds/9071151_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart