CH4102153488 ChemBridge Corporation

(5-chloro-3-methyl-1H-indol-2-yl)methanol

  • MDL number MFCD08691931
  • Cas Number : 77373-72-9
  • Molecular Weight : 195.6455

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071172

Safety Information

MSDS https://www.hit2lead.com/sds/9071172_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart