CH4102153503 ChemBridge Corporation

4-benzoyldihydro-2(3H)-furanone

  • MDL number MFCD00463834
  • Cas Number : 21034-22-0
  • Molecular Weight : 190.1953

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071210

Safety Information

MSDS https://www.hit2lead.com/sds/9071210_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart